Thermo Scientific 88870121 PCR Plate Block for Dry Block Heaters, 96 Well or Full-SkIRt

Representative image only.
Thermo Scientific Mfr # 88870121 – Item # 3640332
Easily switch out blocks for new requirements
  • Get maximum heat-up and retention with close contact of tubes to wall
  • Constructed of solid anodized aluminum for durability
  • Single block dimensions (L x W x H): 5" x 3" x 1 1/2" (12.4 x 7.6 x 3.9 cm).
  • 2-1/2 Years Parts and Labor Warranty

PRODUCT OPTIONS

₹85,200.00INR / Each

SPECS & DESCRIPTION

Specifications & Description

  • Block Number Of Openings96
  • Block Type0.2 mL
  • Block - Tube / Vial Size Accepted (mL)0.2
  • Height (in)17
  • Width (in)4 29/32
  • Depth (in)3
  • DescriptionPCR Plate Block for Dry Block Heaters, 96 Well or Full-SkIRt
  • Warranty2-1/2 Years Parts and Labor
2 and a Half Year Warranty

Key Features

  • Get maximum heat-up and retention with close contact of tubes to wall
  • Constructed of solid anodized aluminum for durability
  • Single block dimensions (L x W x H): 5" x 3" x 1 1/2" (12.4 x 7.6 x 3.9 cm).
  • 2-1/2 Years Parts and Labor Warranty

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat