Thermo Scientific 900011 Electrode fill solution, 4 M KCl with Ag+ 5/cs
Representative image only.
Thermo Scientific Mfr # 900011 – Item # 0573042
Filling solutions for use with pH electrodes and reference electrodes
  • Solutions work best when combined with ionic strength and pH adjusters, which eliminate interferences from other ion species
  • Case of five 60 mL bottles
PRODUCT OPTIONS
₹24,400.00INR / Carton of 5
SPECS & DESCRIPTION
Specifications & Description
  • Ion-Selective ElectrodepH and ISE
Key Features
  • Solutions work best when combined with ionic strength and pH adjusters, which eliminate interferences from other ion species
  • Case of five 60 mL bottles
More About this Item
All filling solutions for reference electrodes come in cartons of one 60-mL or five 50-mL bottles.