Thermo Scientific 9617BNWP Sure-Flow Ion-Selective Electrode; Chloride
Representative image only.
Thermo Scientific Mfr # 9617BNWP – Item # 5882460
Orion-designed for optimum performance and the most comprehensive ISE selection
  • ionplus™ Sure-Flow™ Combination Ion-Selective Electrodes eliminate fumbling and tangles from multiple cables
  • Combination Ion-Selective Electrodes feature both half-cell and reference cell in one electrode
  • Half-Cell Ion-Selective Electrodes are the classic means of taking ion measurements
  • EPA approved for Methods ASTM D512-89 (1999)
  • 3 year warranty
PRODUCT OPTIONS
₹126,000.00INR / Each
Oakton Electrode Storage Solution; 500 mL
₹3,500.00INR / Pint of 1
SPECS & DESCRIPTION
Specifications & Description
  • Electrode TypeSolid-state
  • Reference JunctionSure-flow
  • Reference ElectrolyteOptimum filling solution B
  • Ion-Selective ElectrodeChloride
  • Min pH (pH)2
  • Max pH (pH)12
  • Min Temperature (° C)10
  • Max Temperature (° C)100
  • ConnectorBNC (waterproof when used with Thermo Scientific Orion Star Series meters)
  • DescriptionSure-Flow Ion-Selective Electrode; Chloride
  • Warranty3 year
3 Year Warranty
Key Features
  • ionplus™ Sure-Flow™ Combination Ion-Selective Electrodes eliminate fumbling and tangles from multiple cables
  • Combination Ion-Selective Electrodes feature both half-cell and reference cell in one electrode
  • Half-Cell Ion-Selective Electrodes are the classic means of taking ion measurements
  • EPA approved for Methods ASTM D512-89 (1999)
  • 3 year warranty
More About this Item
Combination Ion-Selective Electrodes are easier to use than reference and half-cells—eliminating fumbling and tangles from multiple cables. Sure-flow® technology provides stable, reproducible results. The easy-to-clean design reduces the risk of inaccurate readings due to clogging of the reference junction. All electrodes include a BNC connector, a 3-ft cable, and manual. Complete systems require-the purchase of a standardizing solution, filling solution for the reference electrode, ionic strength adjustor, and pH adjusters. Chloride, cyanide, Sulfide, nitrate, and fluoride are approved for EPA methods for analysis of wastewater and for NPDES permitting.
Type: Solid state electrode
Concentration range 1.8 to 35,500 ppm
Temperature/pH range: 0 to 80°C/ 2 to 12 pH
Interferences; Cyanide, Bromide, Iodide, Hydoxyl, Sulfide