Thermo Scientific Nunc Surface Binding Assay Microplates, 96-Well, Polystyrene, MaxiSorp® Treated, Round, 0.3 mL; 60/Cs

Representative image only.
Thermo Scientific Nunc Mfr # 449824 – Item # 0192808
Surfaces with different degrees of hydrophilicity—ideal for solid phase immunoassays (ELISA)
  • Engineered for easy/reproducible passive absorption
  • Certified homogeneity of binding
  • Clear polystyrene has excellent optical properties making it ideal for optical measurements
  • Plates are optimized for robotics, plate readers and liquid handlers
  • Alphanumerically labeled rows and columns

PRODUCT OPTIONS

₹120,000.00INR / Case of 60
Often Bought With…
Thermo Scientific™ Nunc™ Microplate Covers
61,200.00 - ₹120,000.00INR / Case of 50

SPECS & DESCRIPTION

Specifications & Description

  • Number of Wells96
  • Well Volume (mL)0.3
  • Min Working Well Volume (mL)0.05
  • Max Working Well Volume (mL)0.25
  • Well Top Diameter (mm)7.1
  • Well ShapeRound
  • Surface TreatmentMaxiSorp
  • Lid (Included)No
  • SterileNo
  • Plate ColorClear
  • MaterialPolystyrene
  • Height (mm)14.5
  • Width (mm)86
  • Length (mm)128

Key Features

  • Engineered for easy/reproducible passive absorption
  • Certified homogeneity of binding
  • Clear polystyrene has excellent optical properties making it ideal for optical measurements
  • Plates are optimized for robotics, plate readers and liquid handlers
  • Alphanumerically labeled rows and columns

More About this Item

Surface binding assay microplate wells facilitate surface adsorption of proteins, antibodies, and other biomolecules. The surface adsorption aids in applications for the basis for quantitative assays such as ELISA.

Choose from treated or untreated plates. Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment. Untreated surfaces are ideal for adding coatings in your lab. MaxiSorp surface treatment is hydrophilic and optimized for binding immunoglobulin but also binds a wide range of other proteins.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat