Thermo Scientific TSX3030FA Auto-Defrost Freezer, 30 cu ft; 120V/60 Hz

Representative image only.
Thermo Scientific Mfr # TSX3030FA – Item # 4420241
Store samples securely—no need to remove when defrosting
  • V-drive technology enhances temperature uniformity by continually adapting to surroundings
  • Whisper quiet operation blends seamlessly into your workspace
  • ENERGY STAR® certified to reduce costs and help the environment
  • Four 2" casters ensures easy mobility; the front two are lockable
  • Self-closing door with 90° stop simplifies inventory loads
  • Capacity is 30 cubic feet
  • Operates on 120 VAC, 60 Hz power
  • 2 Years Parts & Labor Warranty

PRODUCT OPTIONS

Call For Price

Call For Price

Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • Freezer Capacity (cu ft)29.2
  • Freezer Capacity (Liters)827
  • Min Temperature - Freezer (° F)-31
  • Max Temperature - Freezer (° F)5
  • Min Temperature - Freezer (° C)-35
  • Max Temperature - Freezer (° C)-15
  • Temperature ControlMicroprocessor
  • Freezer Interior Width (in)30
  • Freezer Interior Height (in)58
  • Freezer Interior Depth (in)28 1/2
  • Freezer Interior Width (cm)76
  • Freezer Interior Height (cm)147
  • Freezer Interior Depth (cm)72
  • Exterior Width (in)34
  • Exterior Height (in)78 1/2
  • Exterior Depth (in)37 7/8
  • Exterior Width (cm)86
  • Exterior Height (cm)199
  • Exterior Depth (cm)96
  • Number of Doors1
  • Number of Shelves4
  • AlarmYes
  • Power (VAC)115
  • Power (Hz)60
  • DescriptionAuto-Defrost Freezer, 30 cu ft; 120V/60 Hz
  • Warranty2 years parts & labor
2 Year WarrantyUL Listed to Canadian safety standards.Made in the U.S.A.

Includes

USB drive with user manuals, certificate of conformance, control panel keys, cabinet door keys, and power cord.

Key Features

  • V-drive technology enhances temperature uniformity by continually adapting to surroundings
  • Whisper quiet operation blends seamlessly into your workspace
  • ENERGY STAR® certified to reduce costs and help the environment
  • Four 2" casters ensures easy mobility; the front two are lockable
  • Self-closing door with 90° stop simplifies inventory loads
  • Capacity is 30 cubic feet
  • Operates on 120 VAC, 60 Hz power
  • 2 Years Parts & Labor Warranty

More About this Item

Combining quality you can count on with quiet operation to suit your surroundings, this high-performance auto defrost -30°C laboratory freezer is a smart choice, clinically and environmentally. Part of the TSX Series, the ENERGY STAR certified units suit applications requiring storage of reagents, pharmaceuticals, biological and other commonly used laboratory materials such as primers, siRNA molecules and fetal bovine serum (FBS).

No need to remove samples prior to defrosting. These automatic defrost freezers feature forced air circulation for uniform temperature throughout the cabinet, faster door opening recovery and non-invasive temperature- and time-based defrost cycles. Along with unique V-drive technology that optimizes temperature uniformity, value is added by keyed security—for both power switch and setpoint—alarms, ergonomic handles and compatibility with SMART VUE® wireless monitoring systems.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat