Traceable Digital Bottle Thermometer with Calibration; Glass Bead Sensor
Representative image only.
Traceable Item # 9446058
Provides a significant advantage over glass thermometers that can only display current temperature
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Waterproof
  • Displays high/low readings
  • Features a glass bead fill bottle sensor
  • 1 year warranty
PRODUCT OPTIONS
₹8,100.00INR / Each
Time to recalibrate?
for an additional
₹34,000.00INR / Each
for an additional
₹43,100.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Meter Measurement TypeDigital Indicator
  • Number Of Probe Channels1
  • Min Temperature (° F)-4
  • Max Temperature (° F)122
  • Min Temperature (° C)-20
  • Max Temperature (° C)50
  • Accuracy±0.5°C from 0°C to 50°C and ±1°C otherwise
  • Resolution0.1°
  • Battery1 silver oxide lithium (included)
  • Warranty1 year
1 Year WarrantyConformite EuropeenneNIST Traceable Calibration
Key Features
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Waterproof
  • Displays high/low readings
  • Features a glass bead fill bottle sensor
  • 1 year warranty
More About this Item
Accurately monitor temperatures in freezers, refrigerators, incubators, and environmental chambers with this temperature-buffered sensor. The probe is sealed in a miniature bottle filled with nontoxic glycol or glass beads. Bottle and solution are designed to insulate the sensor from rapid changes, such as when a refrigerator door is opened. Easy-to-read digital display shows current temperatures and min/max by monitoring the high and low readings for any time period.
The entire unit is waterproof and comes equipped with a stainless-steel sensor that assures long product life. Supplied with unique chemical-resistant ABS plastic holder that can be placed on a self or affixed to a refrigerator or any wall. Bottle may be removed from holder.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
Guaranteed to have at least one year of calibration remaining on certificate.