Traceable Jumbo Timer
Representative image only.
Traceable Item # 6101826
Effortlessly manage timing with precision and flexibility
  • Simultaneously program and track two independent countdowns with a double-display feature
  • Adjustable jumbo LCD display (90° to 180°) with 1-inch-high digits, readable from 30 feet away
  • Versatile functionality includes count up/down, stopwatch, 12/24-hour clock, and memory recall
  • Loud, distinct alarm signals when time reaches zero, then begins counting up to show elapsed time since alarm
  • 1 year warranty
Loading
₹3,400.00INR / Each
Loading
SPECS & DESCRIPTION
Specifications & Description
  • Timing Capacity23 hours, 59 minutes, 59 seconds
  • Count DirectionUp/down
  • Number of Channels2
  • Resolution1 second
  • Accuracy0.01%
  • AlarmYes
  • DisplayDigital
  • Battery TypeAlkaline
  • Battery IncludedYes
  • Height (in)1/2
  • Depth (in)3 3/4
  • Width (in)5 1/2
  • DescriptionJumbo Timer
  • Warranty1 year
Includes
Traceable calibration certificate, battery, stand, and wall-mount bracket.
More About this Item
The Traceable® Jumbo Timer is designed for ultimate accuracy and convenience, offering precise timing with an accuracy of 0.01% and a resolution of 1 second. Whether mounted on a wall, placed flat, or propped up using the included stand, it adapts to any workspace. The unit’s memory recall simplifies repetitive tasks, while the automatic or manual alarm shutoff ensures flexibility in operation.
Each unit includes a Traceable® Certificate, ensuring accuracy from our ISO/IEC 17025:2017 calibration laboratory accredited by A2LA