Traceable Ultra™ Calibrated Refrigerator/Freezer Thermometer; 1 Bottle and 1 Bullet Probe

Representative image only.
Traceable Item # 9876758
Smart-Alarm™ signals both audible and visual alarms even after return to normal conditions
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Triple display simultaneously shows MIN/MAX and current temperature
  • 10-ft microcable allows refrigerator/freezer doors to close on it
  • Temperature-buffered glycol bottle probe mimics solution
  • Traceable® model number 4734
  • 1 year warranty

PRODUCT OPTIONS

₹13,800.00INR / Each
Time to recalibrate?
for an additional
₹34,000.00INR / Each
for an additional
₹43,100.00INR / Each

SPECS & DESCRIPTION

Specifications & Description

  • Meter Measurement TypeDigital Indicator
  • Number Of Probe Channels1
  • Types Of Probes Accepted1 bottle and 1 Bullet probes
  • Min Temperature (° F)-58
  • Max Temperature (° F)158
  • Min Temperature (° C)-50
  • Max Temperature (° C)70
  • Accuracy±0.5°C (-30 to 50°C); otherwise ±1°C
  • Resolution0.1°
  • AlarmYes
  • Min/Max FunctionYes
  • Low Battery IndicatorYes
  • Display TypeLCD
  • Height (in)4 1/4
  • Width (in)2 3/4
  • Depth (in)3/4
  • No. of Probe(s) Included1
  • Cable Length (in)120
  • Battery2 AAA (included)
  • DescriptionCalibrated Refrigerator/Freezer Thermometer; 1 Bottle and 1 Bullet Probe
  • Warranty1 Year
1 Year WarrantyConformite EuropeenneNIST Traceable Calibration

Includes

glass bottle probe with 10-ft cable, Bullet probe with 10-ft cable, flip-open stand, hook-and-loop strips, magnetic strips, 2 AAA batteries, and NIST-traceable calibration certificate with data.

Key Features

  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Triple display simultaneously shows MIN/MAX and current temperature
  • 10-ft microcable allows refrigerator/freezer doors to close on it
  • Temperature-buffered glycol bottle probe mimics solution
  • Traceable® model number 4734
  • 1 year warranty

More About this Item

Accurately monitor temperatures in refrigerators, freezers, water baths, and incubators. Thermometers have accuracy of ±0.5°C (-30 to 50°C). Built-in audible and visual alarms activate when temperature rises or falls below the alarm set point, and is programmable in 0.1° increments.

Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.

Thermometer comes with bottle and Bullet probes. Bottle probe is filled with nontoxic glycol solution, eliminating the possibility of contamination from mercury. Solution is GRAS (generally recognized as safe) by the FDA. The solution filled bottle simulates the temperature of other stored liquids and eliminates the rapid temperature fluctuation caused by opening the refrigerator/freezer door. Use Bullet probe to measure air and liquid temperatures. Bullet probe sensor and cable may be completely immersed. Sensor measures 3/4"L x 0.187" dia.

Guaranteed to have at least one year of calibration remaining on certificate.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat