TSI 792 VelociCalc Surface Temperature Probe
Representative image only.
TSI Mfr # 792 – Item # 1037240
  • 2 year warranty
PRODUCT OPTIONS
₹56,800.00INR / Each
₹17,000.00 - ₹25,900.00INR / Each
for an additional
₹17,000.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Min Temperature (° F)-40
  • Max Temperature (° F)1200
  • Min Temperature (° C)-40
  • Max Temperature (° C)650
  • Temperature Accuracy±0.1% of reading +2°F (±0.056% of reading +1.1°C)
  • Temperature Resolution0.1°F (0.1°C)
  • DescriptionVelociCalc Surface Temperature Probe
  • Warranty2 year
Two year warrantyNIST Traceable Calibration
Key Features
  • 2 year warranty
More About this Item
Keep your TSI ventilation meter working dependably with these parts and accessories.