TSI 9515 VelociCalc Air Velocity Meter, Remote Probe

Representative image only.
TSI Mfr # 9515 – Item # 1027025
Compact anemometers make airflow measurement easy and effortless
  • Accurate air velocity measurement
  • Integrated probe attachment
  • Easy to read display
  • Model 9525 is intrinsically safe
  • Calibration certificate included
  • 2 year warranty

₹261,000.00INR / Each

₹39,200.00 - ₹49,400.00INR / Each

for an additional
₹39,200.00INR / Each
for an additional
₹49,400.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • Max Air Velocity (ft/min)4000
  • Min Air Velocity (ft/min)0
  • Air Velocity Resolution1 ft/min (0.01 m/s)
  • Air Velocity Accuracy±5% of reading or ±5 ft/min (±0.025 m/s) (whichever is greater)
  • Number Of Temperature Channels1
  • Min Temperature (° F)0
  • Max Temperature (° F)200
  • Min Temperature (° C)-18
  • Max Temperature (° C)93
  • Temperature Accuracy±0.5°F (±0.3°C)
  • Temperature Resolution0.1°F (0.1°C)
  • Data LoggingNo
  • Width (in)3 3/8
  • Height (in)1 3/4
  • Length (in)7
  • Battery4 AA (included)
  • DescriptionVelociCalc Air Velocity Meter, Remote Probe
  • Warranty2 year
Two year warrantyNIST Traceable Calibration

Key Features

  • Accurate air velocity measurement
  • Integrated probe attachment
  • Easy to read display
  • Model 9525 is intrinsically safe
  • Calibration certificate included
  • 2 year warranty

More About this Item

These air velocity meters are ideal for measuring face velocity in fume hoods, spray booths, ventilation systems, or IAQ applications.

Model 10270-25 also measures temperature and features a 40"L (102 cm) telescoping probe designed for use in many applications; ideal for hard to reach areas.

Remote probe for 10270-25 is 40"L fully exteneded (7.6 cm) x 1/4" (0.6 cm) dia tip, with 6 ft (1.8 m) cord

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat