Value Plastics Luer to Hose Barb Fitting, Straight Adapter, White Nylon, Female Luer Thread x 200 Series Barb 5/32" ID; 25/Pk
Representative image only.
Value Plastics Mfr # FTLL240-1 – Item # 5012064
Luer tapers that are constructed to meet ISO 594-1 and 594-2 standards
  • Rotating locks minimize twisting and kinking of tubing during positioning
  • Tapered cone seals are reliable and uncomplicated
  • Facilitate single-use configurations that minimize downtime
PRODUCT OPTIONS
₹2,200.00INR / Pkg of 25
Order your Tubing Today!
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNylon
  • Number of Ports2
  • Connection Type-Port 1Female Luer
  • Connection Type-Port 2Hose Barb
  • Barb Size-Port 2 (in)5/32
  • Regulatory ApprovalsRoHS;Non-DEHP;Non-Phthalate
  • DescriptionLuer to Hose Barb Fitting, Straight Adapter, White Nylon, Female Luer Thread x 200 Series Barb 5/32" ID; 25/Pk
Key Features
  • Rotating locks minimize twisting and kinking of tubing during positioning
  • Tapered cone seals are reliable and uncomplicated
  • Facilitate single-use configurations that minimize downtime
More About this Item
These luer fittings come in many configurations and several materials, joining tubing from 1/16” (1.6 mm) ID to 1/4” (6.4 mm) ID. The taper of the female luer is designed to match the taper of the male luer, creating a secure and leak-free connection when the two parts are joined together. The 200 series barb offers outstanding performance characteristics when used with flexible tubing. Tubing is expanded by 50% above its nominal inside diameter, permitting high-pressure capabilities. Because the fittings are made to ISO standards that are well understood globally, it ensures proper interconnections and often removes the need to retrofit*.
Options include nylon, polycarbonate, or polypropylene resin.
Common features of nylon include a high mechanical strength, great balance of stiffness and toughness, good performance at high temperatures, good electrical and flammability properties, and resistance against abrasion or chemicals. It can be sterilized using gamma radiation, Ethylene oxide (EtO), or autoclave.
Polypropylene is clarified and produced without any animal derived components or phthalates. It can be sterilized using gamma radiation, Ethylene oxide (EtO), E-beam, or autoclave.
Polycarbonate is transparent and features high impact, chemical, UV, and electrical resistance. Sterilization is only recommended via ethylene oxide (EtO). Gamma radiation up to 10 Mrads is possible with minor loss of physical properties – it will discolor to a yellow-green hue. Autoclave is not recommended and may crack the fitting.
*Always test the application to verify the performance of the interconnection.