Weksler Dual Scale Gauge, 1.5" Dial, 30"/0" VAC / 100 to 0 kPa, 1/8" NPT Back Connection
Representative image only.
Weksler Mfr # UA15H8C – Item # 6834514
Quality gauge is designed for general pressure applications
  • Use with automated assembly, agricultural, and process equipment
  • 1 year warranty
PRODUCT OPTIONS
₹1,800.00INR / Each
₹34,000.00 - ₹43,100.00INR / Each
for an additional
₹34,000.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±3-2-3% full-scale (ANSI/ASME Grade B)
  • Dial Size (in)1.5
  • Dial Size (mm)38
  • Max Vacuum (" Hg)30
  • Min Vacuum (" Hg)0
  • Wetted MaterialsBrass, bronze
  • CaseSteel
  • Lens MaterialPlastic
  • Process Connection1/8" NPT(M), back
  • Unit Of Measurementpsi, kPa
  • DisplayAnalog dial
  • Media CompatibilityGases and liquids compatible with wetted materials
  • Warranty1 year
1 Year Warranty
Key Features
  • Use with automated assembly, agricultural, and process equipment
  • 1 year warranty
More About this Item
For your day to day pressure measurement needs, these quality dry gauges are an economical solution for a variety of applications. Gauges feature a white background with kPa in blue on outer scale and psi in black on the inner ring. Case and ring is black finish steel with plastic lens. Case is black steel with a plastic window. Tube is bronze, socket is brass.