WTW PC Software for Easy Data Management, Extended with PhotoLab® Color
Representative image only.
WTW Mfr # 902763 – Item # 1596170
Ideal for wastewater influent and effluent testing, industrial process water testing, and environmental monitoring
  • PhotoLab® 7100 VIS Spectrophotometer covering the range from 320 to 1100 nm. Universal instrument suitable for all routine measurements with test kits, in service labs, industry and R&D, wherever the UV range is t not needed.
  • PhotoLab® 7600 UV-VIS Spectrophotometer covering the range from 190 to 1100 nm.
  • It is Ideal for additional self-control with optical reagent-free methods (OptRF) and UV-VIS procedures for many applications.
PRODUCT OPTIONS
₹52,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionPC Software for Easy Data Management, Extended with PhotoLab® Color
Key Features
  • PhotoLab® 7100 VIS Spectrophotometer covering the range from 320 to 1100 nm. Universal instrument suitable for all routine measurements with test kits, in service labs, industry and R&D, wherever the UV range is t not needed.
  • PhotoLab® 7600 UV-VIS Spectrophotometer covering the range from 190 to 1100 nm.
  • It is Ideal for additional self-control with optical reagent-free methods (OptRF) and UV-VIS procedures for many applications.