WTW pH 3310 ProfiLine meter kit with SenTix 41 pH electrode

Representative image only.
WTW Mfr # 2AA312 – Item # 5889022
A rugged new line of electrochemistry meters with a range of features, benefits, and price points to meet every application and budget!
  • Advanced 3310 models feature an innovative waterproof USB connection for ultra-flexible data output
  • Broad pH buffer recognition to meet most calibration needs
  • Rugged, waterproof housing is great for the lab or the field
  • 3 year warranty

₹304,000.00INR / Each

₹25,500.00 - ₹34,500.00INR / Each

Time to recalibrate? Click on the calibration service to view details, then order.
Oakton Electrode Storage Solution; 500 mL
₹3,500.00INR / Pint of 1
Storage & Cleaning Solutions…
Oakton® pH Electrode Storage and Cleaning Solutions
3,500.00 - ₹67,500.00INR / Case of 24

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypePortable
  • Display typeLCD
  • Min pH (pH)-2
  • Max pH (pH)20
  • pH Resolution0.001/0.01/0.1
  • pH Accuracy±0.005 pH
  • Buffer RecognitionDIN, NIST, and USA
  • CalibrationUp to 5 points
  • Min mV (mV)-2000
  • Max mV (mV)2000
  • mV Resolution0.1 or 1 mV
  • mV Accuracy±0.3 mV + 1 digit
  • Min Temperature (° C)-5
  • Max Temperature (° C)105
  • Min Temperature (° F)23
  • Max Temperature (° F)221
  • Temperature compensationAutomatic or manual
  • InterfaceUSB
  • WaterproofYes
  • Battery4 AA (included)
  • Width (cm)5.595874
  • Height (cm)7.897622
  • Width (in)2 13/64
  • Length (in)7
  • Height (in)3 7/64
  • Warranty3 year
3 Year WarrantyConformite EuropeenneIP 67ISO9001:2000 Certified Supplier

Includes

4 AA batteries.

Key Features

  • Advanced 3310 models feature an innovative waterproof USB connection for ultra-flexible data output
  • Broad pH buffer recognition to meet most calibration needs
  • Rugged, waterproof housing is great for the lab or the field
  • 3 year warranty

More About this Item

Durable meters feature a 100% waterproof and easy-to-clean keypad. Silicone rubber keys provide tactile feedback for error-free operation. The large graphic display allows for excellent readability even under difficult lighting conditions. A built-in calibration timer ensures more reliable results.

pH 3310 meters add extended data memory for logging up to 5000 GLP-compliant data sets. Time-controlled datalogging function offers adjustable data collection intervals—ideal for long-term field monitoring. A special “sleep” mode saves power and extends battery life when performing long-term monitoring. Nonvolatile-memory saves all stored data even if battery power is lost. Data transfer to a PC is made simple via the unique waterproof mini-USB interface.

Meters include: four AA batteries.

Meter kits add: specified probe, calibration solutions, and hard carrrying case.

Meters with drinking water kit further add: Sentix® 81 probe, stand, beaker, measuring vessel, and CD-ROM.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat