WTW SenTix 42 pH Electrode with ATC
Representative image only.
WTW Mfr # 103637 – Item # 5890052
Optimize pH readings
  • Choice of ATC electrode or glass body for chemical compatibility
  • 1 year warranty
PRODUCT OPTIONS
₹81,900.00INR / Each
Oakton Electrode Storage Solution; 500 mL
₹3,500.00INR / Pint of 1
Include Buffers Today!
Oakton® pH Buffer Solution Calibration Packs
5,100.00 - ₹51,000.00INR / Pkg of 3
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionpH Electrode with ATC
  • Warranty1 year
1 Year Warranty
Key Features
  • Choice of ATC electrode or glass body for chemical compatibility
  • 1 year warranty
More About this Item
Replace your pH electrode if it cannot hold calibration, gets dehydrated and cannot be rehydrated, or if it breaks.