2-D Single-Use BioProcess Bags
Readily integrate into your single-use process applications
  • Free of any animal-derived components for greater process purity
  • Manufactured in a Class 10,000 facility
  • Needle-free sampling port for added safety
  • Integrated handle on 2- through 20-L sizes
Replace traditional rigid-walled plastic and glass tubes, flasks, bottles, and carboys with these sterile, single-use liquid handling systems. The collapsible containers minimize the risk of cross-contamination and reduce cleaning and turnaround time. Offering outstanding biocompatibility, chemical compatibility, barrier and strength properties, these bags meet your most demanding bioprocessing applications.
Multilayer film structure is optimized for bioprocess applications. Film materials are consistent throughout the product range; with low-density polyethylene (LDPE) as the fluid contact material and ethyl vinyl alcohol (EVOH) as the gas barrier layer. Products are certified to be free of animal-derived components and pass USP Class VI medical-grade testing.
Bags maintain integrity at temperatures ranging from –80 to 60°, making them suitable for packaging, ambient, and cryogenic storage of a variety of liquids including media, buffers, process liquids, and biological products. Sterilized by gamma radiation, ports are capped and covered, and systems are double-bagged for integrity.
Call For Price
33 variations of this product are available.
See All Product Options Below
33 PRODUCT OPTIONS
Compareitem specifications
Depth (in) Material Width (in)
Availability Pricing
Saint Gobain 2-D Sterile Single-Use BioProcess Bags, 12"L 2-Port; 50 mL, 20/Pk 1594601 Mfr # BP0050MS
COMPARE
Call For Price
Saint Gobain 2-D Sterile Single-Use BioProcess Bags, 12"L 2-Port; 150 mL, 20/Pk 1594602 Mfr # BP0150MS
COMPARE
Call For Price
Saint Gobain 2-D Sterile Single-Use BioProcess Bags, 12"L 2-Port; 500 mL, 20/Pk 1594604 Mfr # BP0500MS
COMPARE
Call For Price
Saint Gobain 2-D Sterile Single-Use BioProcess Bags, 6"L 3-Port; 50 mL, 20/Pk 1594605 Mfr # BP0050M
COMPARE
Call For Price
Saint Gobain 2-D Sterile Single-Use BioProcess Bags, 6"L 3-Port; 150 mL, 20/Pk 1594606 Mfr # BP0150M
COMPARE
Call For Price
Saint Gobain 2-D Sterile Single-Use BioProcess Bags, 6"L 3-Port; 250 mL, 20/Pk 1594607 Mfr # BP0250M
COMPARE
Call For Price
Saint Gobain 2-D Sterile Single-Use BioProcess Bags, 6"L 3-Port; 500 mL, 20/Pk 1594608 Mfr # BP0500M
COMPARE
Call For Price
Saint Gobain 2-D Sterile Single-Use BioProcess Bags, 6"L 3-Port; 1 L, 20/Pk 1594609 Mfr # BP0001L
COMPARE
Call For Price
Saint Gobain 2-D Sterile Single-Use BioProcess Bags, 6"L 3-Port; 2 L, 20/Pk 1594610 Mfr # BP0002L
COMPARE
Call For Price
Saint Gobain 2-D Sterile Single-Use BioProcess Bags, 6"L 3-Port; 3 L, 20/Pk 1594611 Mfr # BP0003L
COMPARE
Call For Price
Saint Gobain 2-D Sterile Single-Use BioProcess Bags, 6"L 3-Port; 5 L, 20/Pk 1594612 Mfr # BP0005L
COMPARE
Call For Price
Saint Gobain 2-D Sterile Single-Use BioProcess Bags, 6"L 3-Port; 10 L, 10/Pk 1594613 Mfr # BP0010L
COMPARE
Call For Price
Saint Gobain 2-D Sterile Single-Use BioProcess Bags, 6"L 3-Port; 20 L, 10/Pk 1594614 Mfr # BP0020L
COMPARE
Call For Price
Saint Gobain BP0050MC Sterile Media Bag, 50 mL, 6" Lead on Ports 1 & 2, 20" on Port 3; 20/Pk 1594615 Mfr # BP0050MC
COMPARE
Call For Price
Saint Gobain BP0150MC Sterile Media Bag, 150 mL, 6" Lead on Ports 1 & 2, 20" on Port 3; 20/Pk 1594616 Mfr # BP0150MC
COMPARE
Call For Price
Saint Gobain BP0250MC Sterile Media Bag, 250 mL, 6" Lead on Ports 1 & 2, 20"on Port 3; 20/Pk 1594617 Mfr # BP0250MC
COMPARE
Call For Price
Saint Gobain 2-D Single-Use BioProcess Bags, 3-Port; 500 mL, 20/Pk 1594618 Mfr # BP0500MC
COMPARE
Call For Price
Saint Gobain BP0001LC Sterile Media Bag, 1 L, 6" Lead on Ports 1 & 2, 20" on Port 3; 20/Pk 1594619 Mfr # BP0001LC
COMPARE
Call For Price
Saint Gobain BP0002LC Sterile Media Bag, 2 L, 6" Lead on Ports 1 & 2, 20" on Port 3; 20/Pk 1594620 Mfr # BP0002LC
COMPARE
Call For Price
Saint Gobain BP0003LC Sterile Media Bag, 3 L, 6" Lead on Ports 1 & 2, 20" on Port 3; 20/Pk 1594621 Mfr # BP0003LC
COMPARE
Call For Price
Showing 1 - 20 of 33