3-D Single-Use BioProcess Bags and Tank Liners

Broad chemical compatibility for safe handling of biopharmaceutical liquids
  • Ideal for cylindrical and rectangular tanks, drums, and plastic totes
  • Free of any animal-derived components for greater process
  • Manufactured in a Class 10,000 facility
  • Needle-free sampling port for added safety
Sterile, single-use bags and liners meet your most demanding bioprocessing applications—they offering outstanding biocompatibility, chemical compatibility, and barrier and strength properties. The collapsible design minimizes the risk of cross-contamination and reduces cleaning and turnaround time. Typical applications include raw material storage, media hydration/preparation, and intermediate or final product hold.
Multilayer film structure is optimized for bioprocess applications. Film materials are consistent throughout the product range; with low-density polyethylene (LDPE) as the fluid contact material, ethyl vinyl alcohol (EVOH) as the gas barrier layer, and a nylon outer layer. Products are certified to be free of animal-derived components and pass USP Class VI medical-grade testing.
Containers maintain integrity at temperatures ranging from –80 to 60°, making them suitable for packaging, ambient, and cryogenic storage of a variety of liquids including media, buffers, process liquids, and biological products. Sterilized by gamma radiation, ports are capped and covered, and systems are double-bagged for integrity.
Choose from three- and four-port bags, from 50 to 500 liters, and either cylindrical or rectangular shape. The four-port bags feature a bottom port to allow for mixing and recirculation through the utilization of a dip tube. One port acts as a needle-free, female luer sampling port, and all others feature MPX-series quick-disconnect insert (male) fittings.
Open liners for cylindrical tanks reduce delays due to unavailability of clean tanks or drums—you save money by cutting labor costs and eliminating the need for expensive equipment or dedicated tanks and drums. Ready for immediate use; sterile and pyrogen-free.
Call For Price

16 variations of this product are available.

See All Product Options Below
16 PRODUCT OPTIONS
Compareitem specifications
Depth (in) Material Width (in)
Availability Pricing
Saint Gobain STERILE CONTAINER 50L 3 PORT 10/PK 1594635 Mfr # BP0050LD
COMPARE
Call For Price
Saint Gobain STERILE CONTAINER 100L 3 PORT 10/PK 1594636 Mfr # BP0100LD
COMPARE
Call For Price
Saint Gobain STERILE CONTAINER 200L 3 PORT 10/PK 1594637 Mfr # BP0200LD
COMPARE
Call For Price
Saint Gobain STERILE CONTAINER 300L 3 PORT 3/PK 1594638 Mfr # BP0300LD
COMPARE
Call For Price
Saint Gobain STERILE CONTAINER 500L 3 PORT 4/PK 1594639 Mfr # BP0500LD
COMPARE
Call For Price
Saint Gobain STERILE CONTAINER 200L 4 PORT 4/PK 1594640 Mfr # BP0200LB
COMPARE
Call For Price
Saint Gobain STERILE CONTAINER 500L 4 PORT 4/PK 1594641 Mfr # BP0500LB
COMPARE
Call For Price
Saint Gobain TANK LINER 20L, sterile, 20/PK 1594642 Mfr # LR0020L
COMPARE
Call For Price
Saint Gobain TANK LINER 30L, sterile, 20/PK 1594643 Mfr # LR0030L
COMPARE
Call For Price
Saint Gobain TANK LINER 40L, sterile, 20/PK 1594644 Mfr # LR0040L
COMPARE
Call For Price
Saint Gobain TANK LINER 50L, sterile, 20/PK 1594645 Mfr # LR0050L
COMPARE
Call For Price
Saint Gobain TANK LINER 100L 20/PK 1594646 Mfr # LR0100L
COMPARE
Call For Price
Saint Gobain TANK LINER 200L 10/PK 1594647 Mfr # LR0200L
COMPARE
Call For Price
Saint Gobain TANK LINER 300L 10/PK 1594648 Mfr # LR0300L
COMPARE
Call For Price
Saint Gobain TANK LINER 400L 10/PK 1594649 Mfr # LR0400L
COMPARE
Call For Price
Saint Gobain TANK LINER 500L 10/PK 1594650 Mfr # LR0500L
COMPARE
Call For Price
Showing 1- 16 of 16

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat