Airfiltronix Ductless Fume Hoods
Customize your hood with a range of filter options
  • Remove vapors and contaminates with many available filters
  • Easily view and access the workspace the clear shield
  • Quiet efficient operation powered by 530 cfm blower
These ductless fume hoods prevent airborne contaminates from escaping into the work environment, ensuring the safety of workers. Contaminated air is drawn through a variety of filters to remove pollutants. Filters are available for both vapors and particulates and they can be combined for a wide range of applications (filters sold separately). The blower and filter assembly is located on top of the enclosure to save valuable bench space. All hoods include a high-efficiency blower with backward curved impeller which moves up to 530 cfm in free air at a noise level of less than 68 dB.
There are three containment hood options to choose from with varying hood widths, suitable for any application.
The straight front ductless fume hoods are constructed of clear, ¼” thick acrylic with polycarbonate panels and durable aluminum framing. These feature a clear vinyl curtain providing easy access while maintaining airflow and containing contaminates.
The sloping front ductless fume hood features an angled clear polycarbonate hinged door which provides easy access and complete visibility to the workspace area. These hoods are constructed of polycarbonate with a durable and easy-to-clean 1/2” thick polyethylene floor.
474,000.00 - ₹845,000.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Exhaust Volume (CFM) Exterior Width (in) Power (VAC)
Availability Pricing
Airfiltronix 200A G24 Ductless Fume Hood, 24" W x 32.5" H; 120 VAC 3370516 Mfr # 200A G24
₹474,000.00INR / Each
COMPARE
530 24 120
Loading
₹474,000.00INR / Each
Airfiltronix 200A G24240V Ductless Fume Hood, 24" W x 32.5" H; 240 VAC 3370517 Mfr # 200A G24240V
₹499,000.00INR / Each
COMPARE
530 24 240
Loading
₹499,000.00INR / Each
Airfiltronix 200A G30 Ductless Fume Hood, 30" W x 32.5" H; 120 VAC 3370518 Mfr # 200A G30
₹523,000.00INR / Each
COMPARE
530 30 120
Loading
₹523,000.00INR / Each
Airfiltronix 200A G30240V Ductless Fume Hood, 30" W x 32.5" H; 240 VAC 3370519 Mfr # 200A G30240V
₹549,000.00INR / Each
COMPARE
530 30 240
Loading
₹549,000.00INR / Each
Airfiltronix 200A G50 Ductless Fume Hood, 50" W x 32.5" H; 120 VAC 3370520 Mfr # 200A G50
₹819,000.00INR / Each
COMPARE
530 50 120
Loading
₹819,000.00INR / Each
Airfiltronix 200A G50240V Ductless Fume Hood, 50" W x 32.5" H; 240 VAC 3370521 Mfr # 200A G50240V
₹845,000.00INR / Each
COMPARE
530 50 240
Loading
₹845,000.00INR / Each
Airfiltronix 200A SL30 Ductless Fume Hood, Sloping Front, 30" W x 66" H; 120 VAC 3370522 Mfr # 200A SL30
₹812,000.00INR / Each
COMPARE
530 30 120
Loading
₹812,000.00INR / Each
Airfiltronix 200A SL30240V Ductless Fume Hood, Sloping Front, 30" W x 66" H; 240 VAC 3370523 Mfr # 200A SL30240V
₹838,000.00INR / Each
COMPARE
530 30 240
Loading
₹838,000.00INR / Each
Showing 1- 8 of 8