Axial Flow Impellers
Axial Flow Impellers (LIGHTNIN® A-100) with SUPERPITCH® design — 1.5 pitch ratio for greater batch mixing capabilities. Standard axial flow; use for wetting dry powders.
37,000.00 - ₹201,000.00INR / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Lightnin 801009PSP 316 SS axial flow impeller ( A-100), 3.6" dia x 5/8" bore dia 0431790 Mfr # 801009PSP
COMPARE
316 SS axial flow impeller ( A-100), 3.6" dia x 5/8" bore dia
₹67,800.00INR / Each
COMPARE
316 SS axial flow impeller ( A-100), 2.7" dia x 3/8" bore dia
₹37,000.00INR / Each
Lightnin 809418 316 SS axial flow impeller ( A-100), 3.1" dia x 5/16" bore dia 0456054 Mfr # 809418
COMPARE
316 SS axial flow impeller ( A-100), 3.1" dia x 5/16" bore dia
₹75,600.00INR / Each
Lightnin 871243 316 SS axial flow impeller ( A-100), 2.7" dia x 5/8" bore dia 5035000 Mfr # 871243
COMPARE
316 SS axial flow impeller ( A-100), 2.7" dia x 5/8" bore dia
₹80,100.00INR / Each
Lightnin 801021PSP 316 SS axial flow impeller ( A-100), 4.5" dia x 5/8" bore dia 5035004 Mfr # 801021PSP
COMPARE
316 SS axial flow impeller ( A-100), 4.5" dia x 5/8" bore dia
₹99,100.00INR / Each
Lightnin 801028PSP 316 SS axial flow impeller ( A-100), 5.2" dia x 5/8" bore dia 5035006 Mfr # 801028PSP
COMPARE
316 SS axial flow impeller ( A-100), 5.2" dia x 5/8" bore dia
₹118,000.00INR / Each
Lightnin 801033PSP 316 SS axial flow impeller ( A-100), 6.0" dia x 5/8" bore dia 5035008 Mfr # 801033PSP
COMPARE
316 SS axial flow impeller ( A-100), 6.0" dia x 5/8" bore dia
₹120,000.00INR / Each
Lightnin 801049PSP 316 SS axial flow impeller ( A-100), 7.7" dia x 5/8" bore dia 5035010 Mfr # 801049PSP
COMPARE
316 SS axial flow impeller ( A-100), 7.7" dia x 5/8" bore dia
₹149,000.00INR / Each
Lightnin 801046PSP 316 SS axial flow impeller ( A-100), 6.9" dia x 3/4" bore dia 5035020 Mfr # 801046PSP
COMPARE
316 SS axial flow impeller ( A-100), 6.9" dia x 3/4" bore dia
₹136,000.00INR / Each
Lightnin 801050PSP 316 SS axial flow impeller ( A-100), 7.7" dia x 3/4" bore dia 5035022 Mfr # 801050PSP
COMPARE
316 SS axial flow impeller ( A-100), 7.7" dia x 3/4" bore dia
₹149,000.00INR / Each
Lightnin 801055PSP 316 SS axial flow impeller ( A-100), 8.9" dia x 3/4" bore dia 5035024 Mfr # 801055PSP
COMPARE
316 SS axial flow impeller ( A-100), 8.9" dia x 3/4" bore dia
₹173,000.00INR / Each
Lightnin 801065PSP 316 SS axial flow impeller ( A-100), 10.0" dia x 3/4" bore dia 5035028 Mfr # 801065PSP
COMPARE
316 SS axial flow impeller ( A-100), 10.0" dia x 3/4" bore dia
₹201,000.00INR / Each
Showing 1- 12 of 12