Babcock and Paley Bottles
Easily measure the percentage of butter fat content in your sample with these specialized bottles. The body of each bottle is a reaction chamber, while the graduated neck is a measuring device. Use the Babcock bottles for measuring liquids and small samples. The Paley bottles feature a large opening on the side of the body to easily accommodate larger or viscous samples. Replacement stoppers are available: please see accessories. All bottles measure 6 1/2" H.
6,500.00 - ₹178,000.00INR / Case of 12
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Overall Height (in) Description
Availability Pricing
DWK Life Sciences (Kimble) Glass Babcock Bottles, Skim Milk Test, 18 gram Sample; 12/cs 3459000 Mfr # 530-50100
COMPARE
6 1/2 Glass Babcock Bottles, Skim Milk Test, 18 gram Sample; 12/cs
₹178,000.00INR / Case of 12
DWK Life Sciences (Kimble) Glass Babcock Bottle, Milk Test, 18 gram Sample; 1/Pack 3459006 Mfr # 1003S-8
COMPARE
6 1/2 Glass Babcock Bottle, Milk Test, 18 gram Sample; 1/Pack
₹8,400.00INR / Each
DWK Life Sciences (Kimble) Glass Babcock Bottles, Ice Cream Test, 18 gram Sample; 12/cs 3459010 Mfr # 1025-10
COMPARE
6 1/2 Glass Babcock Bottles, Ice Cream Test, 18 gram Sample; 12/cs
₹86,900.00INR / Case of 12
DWK Life Sciences (Kimble) Babcock bottle, half & half and ice cream test 3459016 Mfr # 516-20
COMPARE
6 1/2 Babcock bottle, half & half and ice cream test
₹8,600.00INR / Each
DWK Life Sciences (Kimble) Glass Babcock Bottle, Cream/Cheese Test, 9 gram Sample; 1/Pack 3459026 Mfr # 2085S-50
COMPARE
6 1/2 Glass Babcock Bottle, Cream/Cheese Test, 9 gram Sample; 1/Pack
₹6,500.00INR / Each