Binder FD Series Mechanical Convection Ovens
Multifunctional – ideal for optimal uniformity and time-controlled applications
  • Powerful air turbine offers fast and uniform drying of large quantities of samples
  • Outstanding thermal insulation saves operation costs
These forced convection ovens have a specially developed high-performance fan — offering around 20% higher performance than comparable models. Air exchange rate can be adjusted continuously from 0% to 100%.
Ovens are microprocessor controlled, with an LED (Classic) or LCD (Avantgarde) display of time and temperature. Binder ovens feature the Absolutely Precise Temperature (APT.Line) technology that has become the standard in precision temperature chambers — achieve temperature accuracy up to 300° with remarkably short heating and recovery times. An independently adjustable ISO Class 2 safety thermostat with visual alarm provides full protection against chamber overtemperature. These ovens can be stacked (except 9.1 cu ft models) to conserve valuable lab space.
633,000.00 - ₹637,000.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (cu ft) Power (VAC)
Availability Pricing
Binder FD23 Classic Mechanical Convection Oven, 0.7 cu ft, 115 VAC 0501221 Mfr # 9010-0196
COMPARE
0.7 115
₹633,000.00INR / Each
Binder FD23 Classic Mechanical Convection Oven, 0.7 cu ft, 230 VAC 0501222 Mfr # 9010-0194
COMPARE
0.7 230
₹637,000.00INR / Each
Binder FD56 Avantgarde Mechanical Convection Oven, 2.1 cu ft, 230 VAC 0501245 Mfr # 9010-0303
COMPARE
2.1 230
Call For Price
Binder FD56 Avantgarde Mechanical Convection Oven, 2.1 cu ft, 120 VAC 0501259 Mfr # 9010-0304
COMPARE
2.1 120
Call For Price
Binder FD115 Avantgarde Mechanical Convection Oven, 4.1 cu ft, 120 VAC 0501262 Mfr # 9010-0306
COMPARE
4.1 120
Call For Price
Binder FD260 Avantgarde Mechanical Convection Oven, 9.1 cu ft, 230 VAC 0501263 Mfr # 9010-0309
COMPARE
9.1 230
Call For Price
Binder FD260 Avantgarde Mechanical Convection Oven, 9.1 cu ft, 240 VAC 0501264 Mfr # 9010-0310
COMPARE
9.1 240
Call For Price
Showing 1- 7 of 7