Binder Solid.Line Mechanical Convection Ovens
Count on accurate temperature controls
  • Forced air provides excellent temperature uniformity and faster heating time
  • Controller with timer function simplifies customized use
  • Adjustable exhaust air flap accommodates your environment
Enjoy the reliability required in research, quality assurance and other key applications. The APT.line™ preheating chamber technology ensures precise temperatures, while the independently adjustable ISO Class 2 safety thermostat with visual alarm protects against overheating. Allowing for stacking, each oven offers ergonomic door opening and easy handling.
451,000.00 - ₹589,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (cu ft) Power (VAC) Rise Time
Availability Pricing
Binder FD-S 56 Solid.Line Mechanical Convection Oven, 1.9 cu ft; 120V 0501273 Mfr # 9090-0019
₹451,000.00INR / Each
COMPARE
1.9 120 20 mins to 150°C
Loading
₹451,000.00INR / Each
Binder FD-S 56 Solid.Line Mechanical Convection Oven, 1.9 cu ft; 230V 0501274 Mfr # 9090-0018
₹451,000.00INR / Each
COMPARE
1.9 230 20 mins to 150°C
Loading
₹451,000.00INR / Each
Binder FD-S 115 Solid.Line Mechanical Convection Oven, 3.7 cu ft; 120V 0501276 Mfr # 9090-0025
₹589,000.00INR / Each
COMPARE
3.7 120 22 mins to 150°C
Loading
₹589,000.00INR / Each
Binder FD-S 115 Solid.Line Mechanical Convection Oven, 3.7 cu ft; 230V 0501277 Mfr # 9090-0024
₹569,000.00INR / Each
COMPARE
3.7 230 22 mins to 150°C
Loading
₹569,000.00INR / Each