BRAND® Deep-Well Plates
Suitable for storage, motherplates for HTS, cell and tissue culture, and more
  • Choose from 96- and 384-well plates in a variety of volumes and bottoms
  • Polypropylene plates are chemically-resistant, autoclavble @ 121° C
  • Withstand temperatures as low as -80° C
  • Polystyrene plates are highly transparent
The conical V-shaped bottom of plates with 384 wells is ideal for pelleting and maximum sample recovery. Plates with 96 wells feature an edgeless design with U-shape bottoms for optimal sample mixing. One plate, model 07904-31 features a space saving design, requiring 30-percent less space than standard models, and shallower wells with larger cross section, making it ideal for low-profile automated systems.
To reduce the risk of sample evaporation or contamination, deep well sealing mats (sold separately) should be used to provide an effective plug seal for individual wells.
44,800.00 - ₹187,000.00INR / Pkg of 24
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Well Shape Well Volume (mL)
Availability Pricing
BRAND 701350 Deep Well Plate, 96-well, PP, 1.1 mL, Standard, U-Bottom; 24/PK 0790404 Mfr # 701350
COMPARE
Polypropylene Standard, U-Bottom 1.1
₹44,800.00INR / Pkg of 24
BRAND 701352 Deep Well Plate, 96-well, PS, 1.1 mL, Standard, U-Bottom; 32/PK 0790406 Mfr # 701352
COMPARE
Polystyrene Standard, U-Bottom 1.1
₹56,000.00INR / Pkg of 32
BRAND 701354 Deep Well Plate, 96-well, PP, 2.2 mL, Standard, U-Bottom; 24/PK 0790408 Mfr # 701354
COMPARE
Polypropylene Standard, U-Bottom 2.2
₹65,000.00INR / Pkg of 24
BRAND 701340 Deep Well Plate, 96-Well, PP, 1.2 mL, Low-Profile, U-Bottom; 50/PK 0790431 Mfr # 701340
COMPARE
Polypropylene Low-Profile, U-Bottom 1.2
₹88,900.00INR / Pkg of 50
BrandTech 701355 Deep Well Plate, 384-Well, PP, 0.3 mL, V-Bottom; 48/PK 0790433 Mfr # 701355
COMPARE
Polypropylene V-Bottom 0.3
₹187,000.00INR / Pkg of 48