BRAND® Mini Coolers for Microcentrifuge Tubes
Maintain temperature of your samples in microcentrifuge tubes
  • Protect your temperature-sensitive samples
  • Available in three versions with different cooling temperatures
  • Holds 12 microcentrifuge tubes with volumes, ranging from 0.5 to 2.0 mL
  • Constructed of autoclavable polycarbonate
When it comes to maintaining the right frigid temperature, these mini coolers do the job. Simply freeze the cooler and maintain temperatures as low as -70°C for your microcentrifuge tubes.
51,500.00 - ₹52,400.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity Lowest Set Temperature (° C) Tubes Held
Availability Pricing
BRAND 114930 Centrifuge 12-Tube Mini Cooler, PC, 0.5 to 2.0 mL, 0° C for 60 min. 0610139 Mfr # 114930
₹51,500.00INR / Each
COMPARE
0.5 to 2.0 mL microcentrifuge tubes 0 12
Loading
₹51,500.00INR / Each
BRAND 114935 Centrifuge 12-Tube Mini Cooler, PC, 0.5 to 2.0 mL, -20° C for 60 min. 0610141 Mfr # 114935
₹52,400.00INR / Each
COMPARE
0.5 to 2.0 mL microcentrifuge tubes -20 12
Loading
₹52,400.00INR / Each
BRAND 114940 Centrifuge 12-Tube Mini Cooler, PC, 0.5 to 2.0 mL, -70° C for 45 min. 0610142 Mfr # 114940
₹52,400.00INR / Each
COMPARE
0.5 to 2.0 mL microcentrifuge tubes -70 12
Loading
₹52,400.00INR / Each