Cannon Constant-Temperature Baths
Take up to seven viscosity measurements at one time
  • Stainless steel cooling coil lets you operae below or slightly above ambient temperature
  • Temperature selection switch lets you quickly select from 10 popular temperature settings
  • Motor-driven stirrer ensures uniform temperature throughout bath
  • Ideal for kinematic viscosity measurements per ASTM D 445
Control the viscosity of your samples with these Cannon® constant-temperature baths. These unique baths hold up to seven glass capillary viscometers and two thermometers (holders are required—order separately). See-through glass chamber lets you continuously view samples.
All units feature a control circuit that provides proportional temperature control and an accuracy of ±0.01°>C from 20 to 100°>C (±0.03°>C above 100°>C for the high-temperature deluxe models). Select any temperature within the range or select from ten preset temperatures. Fine tuning dial lets you adjust this value to within 0.01°>C of your target temperature. Multiple safety features include overtemperature, low-level, and sensor disconnection cutoffs.
Chamber is made of clear borosilicate glass and includes a reflective stainless steel baffle in the center; the deluxe model also includes two fluorescent lamps on both sides. Housing is constructed of corrosion-resistant epoxy-coated heavy aluminum with insulated stainless steel cover; includes stainless steel cooling coil.
Fill bath with deionized water or oxidation-inhibiting bath oil for high-temperature applications. Order bath oil and other accessories separately, and thermometers from our Temperature section.
Call For Price
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Power (VAC)
Availability Pricing
Cannon 9726-A15 standard constant temperature bath, 240 VAC 9893505 Mfr # 9726-A15
COMPARE
100 230