Casella Personal Air Sampling Pumps
Ensure compliance by tracking amount of time the pump was moving
  • Motion sensor tracks the amount of time pump was moving
  • View all run parameters on full color display
  • Use even in harsh environments - housing is IP65 rated with rubber boot for added protection
  • Won't get in the way - slim, ergonomic design for easier acceptance
  • Transport and analyze samples in lab by attaching a gas bag to rear outlet
Note: All pumps require a docking station for recharging. Select from the single-position or the five-position docking station based on your needs.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Casella Apex2IS Apex2 Standard Air Sampling Pump 8654330 Mfr # Apex2IS
COMPARE
Standard Air Sampling Pump
Call For Price
Casella Apex2 Plus Air Sampling Pump 8654331 Mfr # Apex2ISPlus
COMPARE
Air Sampling Pump w/Bluetooth
Call For Price
Casella Apex2 Pro Air Sampling Pump 8654332 Mfr # Apex2ISPro
COMPARE
Air Sampling Pump w/Programmable Timer