Cecomp® Digital Pressure Gauges with Quad Memory
Gauge''s memory is saved until cleared-even when power is off
  • ±0.25% full-scale accuracy
  • 316 SS wetted parts
  • ABS/polycarbonate NEMA 4X case
  • Zero function
  • Selectable auto-shutoff times
  • Back-lit display
The accuracy for these gauges includes linearity, hysteresis, and repeatability. The three front buttons turn gauge on or off, zeros guage reading, and cycles through functions. The four memories are user-settable of 1,2,3,4 or four wheel designations of LF, RF, LR, and RR. The gauges come factory calibrated and is pass code protected. The shut-off is selectable from 1 minute to 8 hours, and comes factory defaulted to 5 minutes.
122,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Accuracy Wetted Materials Description
Availability Pricing
Cecomp Digital Pressure Gauge with 4 Memory Storage, 1/4" NPT(M); 30" Hg to 200 psi 6892844 Mfr # F20BNBL-15V200PSIG-M4
COMPARE
±0.25% full-scale 316 SS
₹122,000.00INR / Each