CELLTREAT® 50-mL Sterile Reagent Reservoirs, Polystyrene
Ideal for repetitive liquid transfer with multichannel pipettors
  • Reservoirs are presterilized by gamma irradiation
  • V-shaped angled design promotes funneling of liquid to maximize retrieval and reduce waste
  • Constructed of durable white polystyrene for enhanced solution visibility
The pipette reagent reservoirs feature a V-shaped bottom, making multichannel pipetting easier and ensuring complete aspiration of costly reagents. Sterile reservoirs are sterilized by gamma irradiation and come individually wrapped or bulk packed.
42,800.00 - ₹48,300.00INR / Case of 80
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (mL) Package Of Sterile
Availability Pricing
CELLTREAT Scientific Products Reagent Reservoir, Polystyrene, Sterile, 50 mL, Individually Wrapped; 80/Cs 3456758 Mfr # 229290
COMPARE
50 Individually Wrapped Case of 80 Yes
₹42,800.00INR / Case of 80
CELLTREAT Scientific Products Reagent Reservoir, Polystyrene, Sterile, 50 mL, Bulk Packed; 5/Bag, 40 Bags/Cs 3456759 Mfr # 229291
COMPARE
50 Bulk Packed 5/Bag, 40 Bags/Cs Yes
₹48,300.00INR / Case of 200