CELLTREAT® Scientific Products Bottle, Tube, and Flask Caps
Cell culture caps—ideal for aseptic biological reactions
  • Sterilized by gamma irradiation
  • DNase-& RNase-free
  • Non-pyrogenic
These convenient replacement caps for your tubes, bottles, and flask are sterilized by gamma irradiation and ideal for cell culture applications. Choose from solid or vented caps. Solid caps provide a leakproof seal to keep samples secure. Vented caps feature a 0.22 µm hydrophobic membrane allowing for continuous gas exchange.
Choose the cap the is suitable for use with your CELLTREAT® Scientific Products cell culture product. Roller bottles, flasks, and centrifuge tubes sold separately.
13,500.00 - ₹51,600.00INR / Case of 24
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cap Material Cap Style Special Features
Availability Pricing
CELLTREAT Scientific Products Cap for Roller Bottle, Sterile, HDPE, Vented, 0.22 µm; 24/Cs 0183636 Mfr # 229391
COMPARE
HDPE Vented For use with CELLTREAT roller bottles
₹13,500.00INR / Case of 24
CELLTREAT Scientific Products Cap for Erlenmeyer/Fernbach Flasks, Sterile, Polypropylene, 70 mm, Vented, 0.22 µm; 24/Cs 0193183 Mfr # 229895
COMPARE
Polypropylene Vented For use with CELLTREAT erlenmeyer and fernbach flasks
₹51,600.00INR / Case of 24
CELLTREAT Scientific Products Cap for Bio-Reaction Centrifuge Tube, Sterile, HDPE, for 15 mL Tube, Vented, 0.22 µm; 100/Cs 0633642 Mfr # 229473
COMPARE
HDPE Vented For use with CELLTREAT 15 mL bio-reaction centrifuge tubes
₹39,000.00INR / Case of 100
CELLTREAT Scientific Products Cap for Bio-Reaction Centrifuge Tube, Sterile, HDPE, for 50 mL Tube, Vented, 0.22 µm; 100/Cs 0633645 Mfr # 229477
COMPARE
HDPE Vented For use with CELLTREAT 50 mL bio-reaction centrifuge tubes
₹42,500.00INR / Case of 100