CELLTREAT Sterile Bottletop Filters
Large polyethersulfone membranes provide increase flow rates
  • Feature a polyethersulfone membrane—ideal for high flow and low binding applications
  • Available in 0.22 or 0.45 μm pore sizes and membrane diameters of 50, 75, and 90 mm
  • Angled hose barb adapter increases unit stability and accommodates three tubing sizes
  • Use with standard 45-mm threaded solution bottles (bottles not included)
Tapered hosebarb fittings accepts 3/16" (5 mm), 3/8" (10 mm), and 9/16" (15 mm) ID tubing.
47,900.00 - ₹111,000.00INR / Case of 24
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Membrane Material Pore Size (µm)
Availability Pricing
CELLTREAT Scientific Products 229712 Bottletop Filter, 250 mL, .45 µm; 24/cs 0633681 Mfr # 229712
COMPARE
250 Polyethersulfone (PES) 0.45
₹59,500.00INR / Case of 24
CELLTREAT Scientific Products 229713 Bottletop Filter, 500 mL, .45 µm; 24/cs 0633682 Mfr # 229713
COMPARE
500 Polyethersulfone (PES) 0.45
₹73,100.00INR / Case of 24
CELLTREAT Scientific Products 229714 Bottletop Filter, 1000 mL, .45 µm; 24/cs 0633683 Mfr # 229714
COMPARE
1000 Polyethersulfone (PES) 0.45
₹111,000.00INR / Case of 24
CELLTREAT Scientific Products 229715 Bottletop Filter, 150 mL, .22 µm; 24/cs 0633684 Mfr # 229715
COMPARE
150 Polyethersulfone (PES) 0.22
₹47,900.00INR / Case of 24
CELLTREAT Scientific Products 229716 Bottletop Filter, 250 mL, .22 µm; 24/cs 0633685 Mfr # 229716
COMPARE
250 Polyethersulfone (PES) 0.22
₹60,300.00INR / Case of 24
CELLTREAT Scientific Products 229717 Bottletop Filter, 500 mL, .22 µm; 24/cs 0633686 Mfr # 229717
COMPARE
500 Polyethersulfone (PES) 0.22
₹73,500.00INR / Case of 24
CELLTREAT Scientific Products 229718 Bottletop Filter, 1000 mL, .22 µm; 24/cs 0633687 Mfr # 229718
COMPARE
1000 Polyethersulfone (PES) 0.22
₹111,000.00INR / Case of 24
Showing 1- 7 of 7