CELLTREAT Tissue and Cell Culture Roller Bottles, PS
Uniform, hydrophilic surface promotes cell attachment
27,500.00 - ₹57,500.00INR / Case of 24
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Culture Area (cm²) Sterile
Availability Pricing
CELLTREAT Scientific Products 229382 Culture Roller Bottle, nonvented cap, sterile, 490 sq. cm, 24/cs 0183630 Mfr # 229382
COMPARE
1000 490 Yes
₹54,400.00INR / Case of 24
CELLTREAT Scientific Products 229383 Culture Roller Bottle, vented cap, sterile, 490 sq. cm, 24/cs 0183631 Mfr # 229383
COMPARE
1000 490 Yes
₹57,500.00INR / Case of 24
CELLTREAT Scientific Products 229384 Culture Roller Bottle, nonvented cap, sterile, 850 sq. cm,12/CS 0183632 Mfr # 229384
COMPARE
2000 850 Yes
₹30,200.00INR / Case of 12
CELLTREAT Scientific Products 229385 Culture Roller Bottle, vented cap, sterile, 850 sq. cm, 12/cs 0183633 Mfr # 229385
COMPARE
2000 850 Yes
₹32,300.00INR / Case of 12
CELLTREAT Scientific Products 229386 Culture Roller Bottle, nonvented cap, sterile, 1900 sq. cm, 12/cs 0183634 Mfr # 229386
COMPARE
2000 1900 Yes
₹53,100.00INR / Case of 12
CELLTREAT Scientific Products 229387 Culture Roller Bottle, vented cap, sterile, 1900 sq. cm, 12/cs 0183635 Mfr # 229387
COMPARE
2000 1900 Yes
₹55,300.00INR / Case of 12
CELLTREAT Scientific Products 229582 Non-Treated Culture Roller Bottle, 1L, nonvented cap, sterile, 24/cs 0183637 Mfr # 229582
COMPARE
1000 Yes
₹50,400.00INR / Case of 24
CELLTREAT Scientific Products 229584 Non-Treated Culture Roller Bottle, 2L, nonvented cap, sterile, 12/cs 0183638 Mfr # 229584
COMPARE
2000 Yes
₹27,500.00INR / Case of 12
Showing 1- 8 of 8