CELLTREAT Vented Sterile Culture Flasks
Get improved gas exchange via oversized vent area
  • Fernbach style offers a large surface area to volume ratio
Designed for suspension culture applications, these flasks feature 70-mm opening for easy filling and dispensing. Constructed of optically clear, autoclavable polycarbonate. All flasks are supplied sterile (gamma irradiated) and individually double bagged.
86,000.00 - ₹94,400.00INR / Case of 6
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Material Description
Availability Pricing
CELLTREAT Scientific Products 229850 Vented Sterile Erlenmeyer Flasks with Plain Bottom, 2 L, Individually Wrapped; 6/cs 0193176 Mfr # 229850
COMPARE
2200 Polycarbonate Vented Sterile Erlenmeyer Flasks with Plain Bottom, 2 L, Individually Wrapped; 6/cs
₹89,600.00INR / Case of 6
CELLTREAT Scientific Products 229855 Vented Sterile Erlenmeyer Flasks with Baffled Bottom, 2 L, Individually Wrapped; 6/cs 0193177 Mfr # 229855
COMPARE
2400 Polycarbonate Vented Sterile Erlenmeyer Flasks with Baffled Bottom, 2 L, Individually Wrapped; 6/cs
₹90,400.00INR / Case of 6
CELLTREAT Scientific Products 229865 Vented Sterile Fernbach Flasks with Baffled Bottom, 2 L, Individually Wrapped; 6/cs 0193179 Mfr # 229865
COMPARE
2400 Polycarbonate Vented Sterile Fernbach Flasks with Baffled Bottom, 2 L, Individually Wrapped; 6/cs
₹94,400.00INR / Case of 6
CELLTREAT Scientific Products 229870 Vented Sterile Fernbach Flask with Plain Bottom, 3 L, Individually Wrapped; 4/cs 0193180 Mfr # 229870
COMPARE
4000 Polycarbonate Vented Sterile Fernbach Flask with Plain Bottom, 3 L, Individually Wrapped; 4/cs
₹86,000.00INR / Case of 4
CELLTREAT Scientific Products 229875 Vented Sterile Fernbach Flasks with Baffled Bottom, 3L, Individually Wrapped; 6/cs 0193181 Mfr # 229875
COMPARE
4000 Polycarbonate Vented Sterile Fernbach Flasks with Baffled Bottom, 3L, Individually Wrapped; 6/cs
₹86,300.00INR / Case of 6