Centrifuges for Babcock system
These centrifuges are specifically designed for Babcock Testing. All models feature a heater, thermostat (factory set for 140°F (60°C)), thermometer, visiscope, and brakes in a durable cast aluminum housing. The control box on top of the centrifuge provides easy access to the pilot light and motor and heater switches.
Call For Price
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max RCF x g Max Speed (rpm) Description
Availability Pricing
Babcock 220V, 50Hz centrifuge; 8 bottle capacity 1736005 Mfr # 208-220 V, 50 HZ
COMPARE
166 836 Babcock 220V, 50Hz centrifuge; 8 bottle capacity