Chemware Erlenmeyer Perfluoroalkoxy Flasks
Flasks are ideal for ultrapure applications. They can be heated in microwave; maximum temperature is 500degF (260degC). Graduated, chemically inert flasks are transparent for easy viewing of contents.
28,500.00 - ₹63,200.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL)
Availability Pricing
Chemware D1069075 Erlenmeyer Perfluoroalkoxy (PFA) Flask, 100 mL, 1/Pk 0611200 Mfr # D1069075
COMPARE
100
₹28,500.00INR / Each
Chemware D1069077 Erlenmeyer Perfluoroalkoxy (PFA) Flask, 300 mL, 1/Pk 0611210 Mfr # D1069077
COMPARE
300
₹63,200.00INR / Each