Chemware® Graduated PFA Beakers
Chemically inert PFA for the harshest chemicals
  • Easy determine volume with graduations
  • Autoclavable
These PFA beakers are ideal for ultrapure applications. The bottom of the beaker is designed and machined for rapid heat trasnfer. They can be heated in a microwave or directly on a hot plate up to 500°F (260°C). Beakers are transparent for easy viewing, graduated for easy measuring, and have a nonstick, easy-to-clean surface.
16,200.00 - ₹48,400.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
Chemware D1069536 Griffin Style PFA Beaker, 50 mL, 1/Pk 6100110 Mfr # D1069536
COMPARE
50 10 Perfluoroalkoxy (PFA)
₹16,200.00INR / Each
Chemware D1069537 Griffin Style PFA Beaker, 125 mL, 1/Pk 6100120 Mfr # D1069537
COMPARE
125 25 Perfluoroalkoxy (PFA)
₹23,600.00INR / Each
Chemware D1069538 Griffin Style PFA Beaker, 250 mL, 1/Pk 6100130 Mfr # D1069538
COMPARE
250 50 Perfluoroalkoxy (PFA)
₹35,500.00INR / Each
Chemware D1069539 Griffin Style PFA Beaker, 500 mL, 1/Pk 6100140 Mfr # D1069539
COMPARE
500 50 Perfluoroalkoxy (PFA)
₹48,400.00INR / Each