Cole-Parmer® 12-mm Glass Body, General Purpose, Double-Junction pH Electrodes
Save money—just replace electrode
  • Purchase a separate housing and simply replace electrodes as needed Dome bulb offers better protection from breakage Stable in low-ionic strength solutions Offers resistance to sodium ion error Compatible with most systems
General purpose, double-junction electrodes prevent contamination of the AgCl reference cell by solutions that might react with silver (heavy metals, organics, sulfides). Electrodes can be used for measurements up to pH 13. Maximum pressure is 150 psi at 25°C.
25,100.00 - ₹34,900.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
pH Probe, DJ/Glass/1kOhm RTD/12 mm/10'; Spade/BNC
₹30,800.00INR / Each
COMPARE
General Purpose pH electrode, double-junction, 10-ft cable
₹25,100.00INR / Each
COMPARE
pH Probe, DJ/Glass/100Ohm RTD/12 mm/10'; Plug/BNC
₹31,100.00INR / Each
COMPARE
pH Probe, DJ/Glass/100Ohm RTD/12mm/10'; Tinned/BNC
₹34,900.00INR / Each
COMPARE
General Purpose Double-Junction pH Electrode, 25-ft Cable
₹28,500.00INR / Each
COMPARE
pH Probe, DJ/Glass/100Ohm RTD/12mm/25'; Plug/BNC
₹33,300.00INR / Each
COMPARE
pH Probe, DJ/Glass/1kOhm RTD/12mm/25'; Spade/BNC
₹32,400.00INR / Each
Showing 1- 7 of 7