Cole-Parmer® 12-mm Glass Body, High-pH, Double-Junction pH Electrodes
Economical and efficient combination—buy a protective housing assembly, then simply replace the electrode as needed
  • Use for many applications in a variety of housings
High-pH, double-junction electrodes are well suited for pH levels above 13 or high sodium ion concentrations. Double-junction reference protects against contamination from heavy metals, organics, and sulfides. Maximum pressure is 150 psi at 25°>C.
25,100.00 - ₹28,300.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
ATC Element Cable Length (m) Connector
Availability Pricing
COMPARE
3 BNC
₹25,100.00INR / Each
COMPARE
7.6 BNC
₹28,300.00INR / Each