Cole-Parmer® 760 Torr Absolute Digital Reference Gauges, Battery Powered
Replace mercury manometers to elevate precision in your vacuum measurements with portable or continuous-display absolute reference gauges
  • Absolute reference capability for a reading of zero at full vacuum
  • Designed for diverse applications with NEMA 2 protection against splashing water or dirt
  • Portable battery powered models ideal for on-the-go tasks, such as checking vacuum systems and packaging machinery
  • Low voltage powered models cater to continuous display needs, perfect for monitoring vacuum systems and pumps
  • Elevate precision with ±0.25% test gauge accuracy
Cole-Parmer® 760 Torr absolute reference gauges stand out for their accuracy in vacuum measurements at both full vacuum and atmospheric pressure when the gauge port is open. The easy to read, 3-digit LCD display updates at a rate of 3 readings per second.
The battery powered models are meticulously designed for portability, making them well-suited for on-the-go tasks such as checking vacuum systems and packaging machinery. These models provide a practical solution for professionals who require flexibility and mobility in their work, allowing them to easily carry and use the gauge wherever it's needed. On the other hand, our low voltage-powered models are tailored for applications demanding a continuous display of vacuum levels, making them ideal for scenarios like monitoring vacuum systems and pumps. These models offer a reliable, uninterrupted display, ensuring that users have constant access to critical information for efficient operation and maintenance of their vacuum systems. With both options, these gauges provide a versatile and accurate solution for a wide range of industrial and scientific applications.
83,700.00 - ₹98,000.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Accuracy Display Power Source
Availability Pricing
COMPARE
±0.25% FS ±1 LSD, (approx. ±3 torr) 3 digit LCD, 0.5" H digits Battery
₹83,700.00INR / Each
COMPARE
±0.25% FS ±1 LSD, (approx. ±3 torr) 3 digit LCD, 0.5" H digits Battery
₹83,700.00INR / Each
COMPARE
±0.25% FS ±1 LSD, (approx. ±3 torr) 3 digit LCD, 0.5" H digits Battery
₹83,700.00INR / Each
COMPARE
±0.25% FS ±1 LSD, (approx. ±3 torr) 3 digit LCD, 0.5" H digits Battery
₹83,700.00INR / Each
COMPARE
±0.25% FS ±1 LSD, (approx. ±3 torr) 3 digit LCD, 0.5" H digits, LED backlight Battery
₹90,500.00INR / Each
COMPARE
±0.25% FS ±1 LSD, (approx. ±3 torr) 3 digit LCD, 0.5" H digits, LED backlight Battery
₹90,500.00INR / Each
COMPARE
±0.25% FS ±1 LSD, (approx. ±3 torr) 3 digit LCD, 0.5" H digits, LED backlight Battery
₹90,500.00INR / Each
COMPARE
±0.25% FS ±1 LSD, (approx. ±3 torr) 3 digit LCD, 0.5" H digits, LED backlight Battery
₹98,000.00INR / Each
Showing 1- 8 of 8