Cole-Parmer® BioSafe Serological Pipettes
Filtered for sample integrity and user safety
  • Eliminate cross-contamination — unique barrier filter prevents over-aspiration
  • Polystyrene pipettes are sterile, individually wrapped, and disposable
  • Ideal for use with potentially hazardous or dangerous biological fluids
  • Use in place of filtered pipette tips
These polystyrene serological pipettes feature a special filter plug to lock in fluid and vapors if over aspiration occurs. They also protect electronic pipettors from contamination and cross-contamination of samples from over-aspiration — promoting user safety and sample integrity.
5,600.00 - ₹8,300.00INR / Case of 200
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Graduation Interval Max Volume (mL)
Availability Pricing
COMPARE
0.1 mL 5
₹5,600.00INR / Case of 200
COMPARE
0.1 mL 10
₹7,200.00INR / Case of 200
COMPARE
0.2 mL 25
₹8,300.00INR / Case of 150