Cole-Parmer® CELLroll Cell Cultivation Systems
Easy to install, just click and roll
  • Ideal for vaccine, insect, and antibody cell culture production
  • Advance remote functions and modular design allow easy cell culture scale-up
CELLROLL systems are programmable, modular roller systems that accommodate roller bottles with culture volumes from 300 to 400 mL. Systems operate by remote control and feature two ranges of rotating speeds. The control panel features a digital display, timer, and alarm; program for direction of rotation, bottle angle, velocity, and interval cycles. Systems are CO2 and moisture resistant.
Systems include two decks that hold four roller bottles and can be expanded to accommodate up to 16 bottles (total of eight decks). Fits vertically or horizontally inside a CO2 incubator. Control unit can be used outside the incubator for simple and flexible programming. One deck measures 11-5/8"L x14-3/16"W x 9-¼"H.
1,308,000.00 - ₹1,385,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Power (VAC) Description
Availability Pricing
COMPARE
400 90 to 260 CELLROLL system, 0.1 to 2.0 rpm, 90-260 VAC
₹1,385,000.00INR / Each
COMPARE
400 90 to 260 CELLROLL system, 2.0 to 6.0 rpm, 90-260 VAC
₹1,308,000.00INR / Each