Cole-Parmer® Cleanroom Hose Barb Fittings, Reducing Elbow, Animal-Derivative-Free Polypropylene
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Quick and easy assembly without tools
  • Single barb design tightly “grips” the tubing
  • Meets FDA requirements 21 CFR 177.1520(a)(3)(i) and (c)3.1a and RoHS compliant
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
The reducing elbows feature a 90° bend and is designed to fit in a tight space. A single barb on each end of the fitting creates a tight seal to keep the tubing in place. These animal-derivative-free polypropylene fittings are radiation stable and resistant to solvents, chemicals, water, strong mineral acids, bases, and other inorganic environments. Not recommended for use with oxidizing agents.
1,700.00 - ₹7,200.00INR / Pkg of 10
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Connection Type
Availability Pricing
COMPARE
3/32 1/16 Hose Barb Reducer
₹1,700.00INR / Pkg of 10
COMPARE
1 3/4 Hose Barb Reducer
₹7,200.00INR / Pkg of 10
COMPARE
1/8 3/32 Hose Barb Reducer
₹2,100.00INR / Pkg of 10
COMPARE
1/8 1/16 Hose Barb Reducer
₹2,100.00INR / Pkg of 10
COMPARE
3/16 1/8 Hose Barb Reducer
₹3,300.00INR / Pkg of 10
COMPARE
3/8 1/4 Hose Barb Reducer
₹1,800.00INR / Pkg of 10
COMPARE
1/2 1/4 Hose Barb Reducer
₹1,800.00INR / Pkg of 10
COMPARE
1/2 3/8 Hose Barb Reducer
₹1,800.00INR / Pkg of 10
Showing 1- 8 of 8