Cole-Parmer® Cleanroom Hose Barb Fittings, Straight Union, Animal-Derivative-Free Polypropylene
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Quick and easy assembly without tools
  • Single barb design tightly “grips” the tubing
  • Meets FDA requirements 21 CFR 177.1520(a)(3)(i) and (c)3.1a and RoHS compliant
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
Easily connect two tubing with straight unions. A single barb on each end of the fitting creates a tight seal to keep the tubing in place. These animal-derivative-free polypropylene fittings are radiation stable and resistant to solvents, chemicals, water, strong mineral acids, bases, and other inorganic environments. Not recommended for use with oxidizing agents.
1,100.00 - ₹2,600.00INR / Pkg of 10
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Fitting Material
Availability Pricing
COMPARE
3/32 3/32 Polypropylene
₹1,100.00INR / Pkg of 10
COMPARE
5/8 5/8 Polypropylene
₹1,800.00INR / Pkg of 10
COMPARE
1/16 1/16 Polypropylene
₹1,100.00INR / Pkg of 10
COMPARE
3/4 3/4 Polypropylene
₹2,600.00INR / Pkg of 10
COMPARE
1 1 Polypropylene
₹2,300.00INR / Pkg of 10
COMPARE
1 1 Polypropylene
₹2,600.00INR / Pkg of 10
COMPARE
5/32 5/32 Polypropylene
₹1,800.00INR / Pkg of 10
COMPARE
1/8 1/8 Polypropylene
₹1,400.00INR / Pkg of 10
COMPARE
3/16 3/16 Polypropylene
₹1,800.00INR / Pkg of 10
COMPARE
1/4 1/4 Polypropylene
₹1,300.00INR / Pkg of 10
COMPARE
5/16 5/16 Polypropylene
₹1,800.00INR / Pkg of 10
COMPARE
3/8 3/8 Polypropylene
₹1,300.00INR / Pkg of 10
COMPARE
1/2 1/2 Polypropylene
₹2,000.00INR / Pkg of 10
Showing 1- 13 of 13