Cole-Parmer® Cleanroom Swabs
Ideal for cleaning small surface areas
  • Ultralow non-volatile residue Low particle and fiber generation Good sorbency No contaminating adhesives Excellent chemical resistance
Cole-Parmer cleanroom double-layer polyester swab tip with long polypropylene handle is an excellent general-purpose swab for cleaning surfaces and hard-to-reach areas.
Its laundered, double-layer polyester tip is extremely clean. A long handle and flexible internal head paddle provide good support. Widely recommended for surface sampling and cleaning-validation work.
Features:
• Cleanroom-laundered, double-layer polyester tip over flexible paddle
• Long, light green polypropylene handle
• Thermally bonded head
• Soft and nonabrasive
Applications:
• Cleaning with solvents such as IPA
• Applying lubricants and other liquids
• Removing excess materials
• Sampling surfaces for cleaning validation
• Environmental monitoring for surface bioburden
13,200.00 - ₹60,700.00INR / Case of 1000
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Swab Total Length (InchShort) Tip Length (InchShort) Description
Availability Pricing
COMPARE
6 25/64 21/32 Cleanroom Swab, 0.661"L Polyester Swab Head, PP Handle, Non Sterile; 1000 Pcs/Cs
₹13,200.00INR / Case of 1000
COMPARE
6 25/64 21/32 Cleanroom Swab, 0.661"L Polyester Swab Head, PP Handle, Sterile; 1000 Pcs/Cs
₹17,700.00INR / Case of 1000
COMPARE
6 25/64 21/32 Cleanroom Swab, 0.661"L Polyester Swab Head, PP Handle, Non Sterile; 1500 Pcs/Cs
₹55,200.00INR / Case of 1500
COMPARE
6 25/64 21/32 Cleanroom Swab, 0.661"L Polyester Swab Head, PP Handle, Sterile; 1500 Pcs/Cs
₹60,700.00INR / Case of 1500
COMPARE
6 25/64 21/32 Cleanroom Swab, 0.661"L Polyester Swab Head, PP Handle, Non Sterile; 400 Pcs/Cs
₹53,000.00INR / Case of 400
COMPARE
6 25/64 21/32 Cleanroom Swab, 0.661"L Polyester Swab Head, PP Handle, Sterile; 400 Pcs/Cs
₹55,900.00INR / Case of 400