Cole-Parmer® Crimp Cap for Vials
Aluminum crimps tightly around neck for a tight seal
  • Select from a variety of caps and septa materials that fit your application and needs
  • Magnetic bimetallic caps available
Constructed of aluminum, these crimp vials are ideal for use with your crimp neck vials. To ensure autosamplers function reliably, these caps and septa meet the rigorous specifications demanded by both the analysis and the instrument itself. Defined hardness and cleanliness properties ensure there is a septum available to be both fully compatible with the autosampler needle and meet the demands of the specific analysis.
5,200.00 - ₹178,000.00INR / Pkg of 1000
55 variations of this product are available.
See All Product Options Below
55 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cap color Cap Diameter (mm) Septum Type
Availability Pricing
COMPARE
Silver 20 Pharma-Fix Butyl/PTFE
₹80,900.00INR / Pkg of 1000
COMPARE
Silver 20 White Silicone/PTFE Septa
₹108,000.00INR / Pkg of 1000
COMPARE
Silver 20 Blue Transparent Silicone/PTFE Septa
₹117,000.00INR / Pkg of 1000
COMPARE
Silver 20 Dark Red Silicone/PTFE
₹178,000.00INR / Pkg of 1000
COMPARE
Silver 20 Butyl/PTFE Septa
₹57,700.00INR / Pkg of 1000
COMPARE
Silver 20 White Silicone/Aluminum Foil
₹150,000.00INR / Pkg of 1000
COMPARE
Silver 8 Rubber/PTFE
₹11,200.00INR / Pkg of 1000
COMPARE
Silver 8 PTFE/Silicone/PTFE
₹14,800.00INR / Pkg of 1000
COMPARE
Silver 8 Silicone/Red PTFE
₹13,200.00INR / Pkg of 1000
COMPARE
Silver 8 Pre-Slit Silicone/Red PTFE
₹62,900.00INR / Pkg of 1000
COMPARE
Silver 8 Silicone/White PTFE
₹13,000.00INR / Pkg of 1000
COMPARE
Silver 11 Rubber/PTFE
₹13,700.00INR / Pkg of 1000
COMPARE
Blue 11 Rubber/Butyl/PTFE
₹16,000.00INR / Pkg of 1000
COMPARE
Silver 11 Rubber/Butyl/PTFE
₹19,300.00INR / Pkg of 1000
COMPARE
Silver 11 Rubber/PTFE
₹11,800.00INR / Pkg of 1000
COMPARE
Green 11 Rubber/Butyl/PTFE
₹16,000.00INR / Pkg of 1000
COMPARE
Red 11 Rubber/Butyl/PTFE
₹16,000.00INR / Pkg of 1000
COMPARE
Red 11 Rubber/PTFE
₹24,900.00INR / Pkg of 1000
COMPARE
Gold 11 Rubber/Butyl/PTFE
₹16,000.00INR / Pkg of 1000
COMPARE
Silver 11 PTFE/Silicone/PTFE
₹23,700.00INR / Pkg of 1000
Showing 1 - 20 of 55