Cole-Parmer® DC-400 Automatic Desiccator Cabinets
Automated dehumidification system offers convenience
  • Set relative humidity range (RH) between 25 and 55%
  • Built-in digital hygrometer allows easy monitoring of RH
  • View your samples through acrylic viewing panels
Keeping a constant RH level has never been easier. The electronic dehumidification system includes a permanent desiccant and a hygrometer to keep the relative humidity level between 25 and 55%. The desiccant automatically recycles by discharging absorbed moisture through an external shutter. Once it is recycled, desiccant is ready to absorb moisture again. Safely keep humidity sensitive products such as optical equipment, electronic parts, PCB boards, and reagents.
Aluminum frame and stainless steel (SS) shelves offer excellent durability. SS shelves have holes to ensure uniform humidity throughout chamber. A lock and key let you securely lock the cabinet to prevent unauthorized access.
240,000.00 - ₹255,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Chamber Volume (Liters) Exterior Height (InchShort) Power (VAC)
Availability Pricing
Cole-Parmer DC-400 Automatic Desiccator Cabinet, 25 to 55% RH, One Shelf; 50 L 7890285
₹240,000.00INR / Each
COMPARE
50 17 110
Loading
₹240,000.00INR / Each
Cole-Parmer DC-400 Automatic Desiccator Cabinet, 25 to 50% RH, Two Shelves; 70 L 7890287
₹255,000.00INR / Each
COMPARE
70 22.9 110
Loading
₹255,000.00INR / Each