Cole-Parmer® Differential pH Meter Accessories
Accessories for the Cole-Parmer pH and ORP differential sensors
  • Quick and easy maintenance with optional replacement salt bridge
Sensor life is extended with the replacement salt bridge feature and refill solution, which avoids having to replace the entire sensor. Choose from the replacement salt bridge in packs of either 1, 3, 10 10 each. The standard cell refill solution is available in three sizes as well.
9,300.00 - ₹95,600.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Salt bridge for SD7000CD &-ORP, 1 each
₹11,900.00INR / Each
COMPARE
Salt bridge for SD7000CD & -ORP, 3 each
₹28,400.00INR / Pkg of 3
COMPARE
Salt bridge for SD7000CD & -ORP, 10 each
₹95,600.00INR / Pkg of 10
COMPARE
Standard cell refill solution, 125 mL
₹9,300.00INR / Each