Cole-Parmer® Disposable Lab Containers, PPCO & HDPE
Resists damage from boiling and freezing
  • Impervious to most lab chemicals (including formaldehyde)
Polypropylene copolymer (PPCO) and HDPE lab containers are stackable, translucent, and come with tight-fitting linear low density polyethylene (LLDPE) lids.
22,100.00 - ₹44,800.00INR / Box of 100
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Material Description
Availability Pricing
COMPARE
120 Polypropylene Copolymer Disposable Lab Container, Polypropylene Copolymer (PPCO), 4 oz
₹44,700.00INR / Box of 250
COMPARE
240 Polypropylene Copolymer Disposable Lab Container, Polypropylene Copolymer (PPCO), 8 oz
₹43,600.00INR / Box of 250
COMPARE
480 HDPE Disposable Lab Container, HDPE, 16 oz
₹28,000.00INR / Box of 100
COMPARE
960 Polypropylene Copolymer Disposable Lab Container, Polypropylene Copolymer (PPCO), 32 oz
₹44,800.00INR / Box of 100
COMPARE
2000 Polypropylene Copolymer Disposable Lab Container, Polypropylene Copolymer (PPCO), 2 Qt.
₹35,200.00INR / Box of 50
COMPARE
2500 Polypropylene Copolymer Disposable Lab Container, Polypropylene Copolymer (PPCO), 2-1/2 Qt.
₹22,100.00INR / Box of 25
COMPARE
5000 HDPE Disposable Lab Container, HDPE, 5-1/4 Qt.
₹34,900.00INR / Box of 25
Showing 1- 7 of 7