Cole-Parmer® Disposable Micro Spatulas, Sterile, PS
Dispense and sample small quantities of powders and granules
  • Manufactured and packed in an ISO Class 8 (Class 100,000) cleanroom to ensure cleanliness and reduce cross-contamination
  • Made from high-impact polystyrene (HIPS) which conforms to FDA, EC, and EU regulations
  • BSE/TSE free
The single-use spatulas are gamma irradiated and individually packaged so they can be used straight out of the bag without cleaning.
38,200.00 - ₹40,200.00INR / Pkg of 100
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Overall Length (cm) Width (cm) Description
Availability Pricing
Cole-Parmer Disposable Flat Spatulas, Sterile, PS; 100/PK 0626420
₹40,200.00INR / Pkg of 100
COMPARE
17.2 1.3 Disposable Flat Spatulas, Sterile, PS; 100/PK
Loading
₹40,200.00INR / Pkg of 100
Cole-Parmer Disposable Micro Spatulas, Sterile, PS, 2 mL; 100/PK 0626421
₹38,200.00INR / Pkg of 100
COMPARE
17.5 1.3 Disposable Micro Spatulas, Sterile, PS, 2 mL; 100/PK
Loading
₹38,200.00INR / Pkg of 100
Cole-Parmer Disposable Micro Spatulas, Very Narrow, Sterile, PS, 0.3 mL; 100/PK 0626422
₹38,200.00INR / Pkg of 100
COMPARE
17 0.4 Disposable Micro Spatulas, Very Narrow, Sterile, PS, 0.3 mL; 100/PK
Loading
₹38,200.00INR / Pkg of 100
Cole-Parmer Disposable Micro Spatulas, Narrow, Sterile, PS, 0.5 mL; 100/PK 0626423
₹38,200.00INR / Pkg of 100
COMPARE
17 0.6 Disposable Micro Spatulas, Narrow, Sterile, PS, 0.5 mL; 100/PK
Loading
₹38,200.00INR / Pkg of 100