Cole-Parmer® Disposable Plastic Serological Pipettes
Filter plug prevents overfilling
  • Color coded rings ease identification
  • Constructed from transparent medical-grade polystyrene
  • Sterilized by gamma irradiation and tested before lots are released
Designed for accurate and rapid fluid dispensing, these disposable polystyrene (PS) serological pipettes feature a filter plug which eliminates lint fibers and a uniform mouth piece. Sharp, black, permanent graduations guarantee precise reading and accurate dispensing to ±2%. Reverse graduations show volume removed or remaining on all pipettes larger than 1 mL. Negative graduations provide additional pipetting volume.
Pipette with 1.1-mL capacity meets the APHA standards for the examination of dairy products.
Individually wrapped pipettes come in a fiber-free paper with easy peel-apart opening. Bulk pipettes come in packs of 25. Color-coded dispenser box allows quick access to pipettes without opening top.
9,200.00 - ₹24,000.00INR / Case of 50
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (mL) Description
Availability Pricing
COMPARE
1.1 Disposable Serological Pipettes, 1.1 mL, Milk, 40 Pks of 25; 1000/CS
₹20,300.00INR / Case of 1000
COMPARE
1.1 Disposable Serological Pipettes, 1.1 mL, Milk, Individually Wrapped; 500/Cs
₹9,200.00INR / Case of 500
COMPARE
2.2 Disposable Serological Pipettes, 2.2 mL, Individually Wrapped; 500/CS
₹13,600.00INR / Case of 500
COMPARE
2 Disposable Serological Pipettes, 2 mL, 40 Pks of 25; 1000/CS
₹24,000.00INR / Case of 1000
COMPARE
10 Disposable Serological Pipettes, 10 mL, Individually Wrapped; 200/CS
₹9,400.00INR / Case of 200
COMPARE
100 Disposable Serological Pipettes, 100 mL, Individually Wrapped; 50/CS
₹12,000.00INR / Case of 50