Cole-Parmer® Economical, In-Line ORP electrodes
Integral, one piece electrode and housing for installation convenience
  • Choose from single-junchtion or double-junction designs
  • Available with BNC or tinned ends
In-line electrodes operate from 23 to 212°>F (-5 to 100°>C) and have a chemical-resistant PVDF housing with a 1/2'' NPT(M) thread. Housing measures 5'' x 15/16'' diameter. Maximum pressure 100 psi. Electrode extends 1'' beyond housing.
36,600.00 - ₹36,700.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
ORP electrode, in-line, single-junction, 10-ft cable, BNC
₹36,600.00INR / Each
COMPARE
ORP electrode, in-line, double-junction, 10-ft cable, BNC
₹36,700.00INR / Each